missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min

Manufacturer:  Alfa Aesar™ 1255206

 View more versions of this product

Catalog No. AA1255206



Specifications

Specifications

Gold(I) potassium cyanide
(metals basis)
MFCD00011414
6235525
Potassium dicyanoaurate(I); Potassium gold cyanide
288.33
99.96%
Au 67.6% min.
Odorless
(decomposition)
13967-50-5
KAu(CN)2
UN1588
14,7628
Soluble in water and slightly soluble in alcohol.
Crystalline Powder
Premion™
3.45 g/mL
White
5g

Chemical Identifiers

13967-50-5
MFCD00011414
Potassium dicyanoaurate(I); Potassium gold cyanide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.

P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P390-P501c

H290-H300+H310+H330-H315-H318

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 237-748-4

RTECSNumber : CK6270000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents