missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold(I) iodide, 99%

Manufacturer:  Alfa Aesar™ 1661703

 View more versions of this product

Catalog No. AA1661703



Specifications

Specifications

Gold(I) iodide
AuI
UN3260
Insoluble in water; Soluble in alkali iodide, cyanide solutions; Decomposes in warm acids
I[Au]
323.871
323.87
99%
Moisture Sensitive
1g
10294-31-2
MFCD00014175
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
ATGIETUGWDAYPU-UHFFFAOYSA-M
iodogold
82526
Crystalline
8.25g/mL
120°C (decomposition)

Chemical Identifiers

10294-31-2
323.871
ATGIETUGWDAYPU-UHFFFAOYSA-M
82526
I[Au]
AuI
MFCD00014175
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
iodogold
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents