Please login to your online account to display your discounted pricing

Gold(I) cyanide, Yellow powder, Premion™, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Gold(I) cyanide
CAS 506-65-0
Physical Form Powder
Assay Percent Range 99.99% (Metals basis)
Molecular Formula AuCN
Formula Weight 222.98
MDL Number MFCD00003437
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3203101

  SDS
alfa aesar
3203101
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3203103

  SDS
alfa aesar
3203103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3203106

  SDS
alfa aesar
3203106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 222.98
Quantity 0.25g
Color Yellow
DOT Information DOT Class: 6.1, Packing Group: II
Sensitivity Moisture sensitive
Assay Percent Range 99.99% (Metals basis)
Molecular Formula AuCN
Health Hazard 1 H301-H310-H330-H315-H318-H290-H400-H410: Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Chemical Name or Material Gold(I) cyanide
EINECS Number 208-049-1
Odor Almond-like
MDL Number MFCD00003437
CAS 506-65-0
Health Hazard 2 Keep container tightly closed. Do not empty into drains. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Safety and Handling

H301-H310-H330-H315-H318-H290-H400-H410: Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

Keep container tightly closed. Do not empty into drains. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.