Please login to your online account to display your discounted pricing

Gold(I) cyanide, ACROS Organics™

$291.29 - $291.29

Chemical Identifiers

Chemical Name or Material Gold(I) cyanide
CAS 506-65-0
Molecular Formula AuCN
Linear Formula AuCN
Formula Weight 222.98
MDL Number MFCD00003437
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390132500

  SDS
thermo fisher scientific
AC390132500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390130010

  SDS
thermo fisher scientific
AC390130010
Glass bottle 1g Each for $291.29
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 222.98
Quantity 250mg
Color Yellow
Merck Index 15, 4551
Linear Formula AuCN
MDL Number MFCD00003437
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Gold(I) cyanide
Gold (Au) 87.0 min.
Packaging Glass bottle
Molecular Formula AuCN
CAS 506-65-0
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.