Please login to your online account to display your discounted pricing

Gold(I) cyanide, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC390130010

 View more versions of this product

Catalog No. AC390130010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 222.98
Physical Form Powder
Quantity 1g
Color Yellow
Merck Index 15, 4551
Chemical Name or Material Gold(I) cyanide
CAS 506-65-0
Gold (Au) 87.0 min.
Molecular Formula AuCN
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
MDL Number MFCD00003437
Packaging Glass bottle
Linear Formula AuCN
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Gold(I) cyanide
CAS 506-65-0
Molecular Formula AuCN
Linear Formula AuCN
Formula Weight 222.98
MDL Number MFCD00003437
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.

SDS & Safety Documents