Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold Germanium slug, 6.35mm (0.25in)dia x 6.35mm (0.25 in.) length, Premion™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold Germanium slug
Name Note 6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
Purity Grade Premion™
MDL Number MFCD02091706
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14173KG

  SDS
alfa aesar™
14173KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14173KJ

  SDS
alfa aesar™
14173KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Slug
Chemical Name or Material Gold Germanium slug
Name Note 6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
Quantity 1 piece
MDL Number MFCD02091706
Purity Grade Premion™
TSCA TSCA
Odor Odorless
Weight ≈2.9g/slug
Assay Percent Notes (metals basis)
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Assay 99.999%