Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold Germanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Premion™, 99.9997% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold Germanium slug
Name Note 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Purity Grade Premion™
MDL Number MFCD02091706
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14172KG

  SDS
alfa aesar™
14172KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14172KJ

  SDS
alfa aesar™
14172KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold Germanium slug
Odor Odorless
Physical Form Slug
MDL Number MFCD02091706
Weight ≈2.9g/slug
Name Note 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Purity Grade Premion™
TSCA TSCA
Quantity 1 piece
Assay Percent Notes (metals basis)
Assay 99.9997%
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%