missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals
Form: Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.
Supplier: Thermo Scientific Chemicals 040930FI
Description
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Golden | |
7440-57-5 | |
99.99% (Metals basis) | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
52 Mesh | |
Wire Cloth |
Gold | |
Gold colored gauze | |
Au | |
50 x 50 mm | |
Soluble in aqua regia. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Odorless |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only