missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold foil, >99.9975% (metals basis), Alpha Aesar, Premion™

Form: approx. 1.2g/25x25mm

Manufacturer:  Alfa Aesar 11391GH

 View more versions of this product

Catalog No. AA11391GH



Specifications

Specifications

Gold
7440-57-5
>99.9975% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Golden
100 x 100mm
Gold colored foil
Au
burnish
[Au]
Odorless
23985
Foil, 0.1mm (0.004 in.) thick

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

Documents