missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold foil, 99.95% (metal basis), Alfa Aesar™

Form: approx. 1.2g/25x25mm

Manufacturer:  Alfa Aesar 14721FF

 View more versions of this product

Catalog No. AA14721FF



Specifications

Specifications

Gold
7440-57-5
19.32g/cm3 at 20°C
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
Odorless
gold
23985
Wire, 0.5mm (0.02 in.) dia.
25 x 25mm
Gold colored wire
99.999% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

SDS Documents