missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold foil, 2.0mm (0.08 in.) thick, Premion™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 44166FI

 View more versions of this product

Catalog No. AA44166FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gold foil
50 x 50mm
7440-57-5
Au
burnish
[Au]
Odorless
23985
Foil
Premion™
≈24g/25x25mm
2.0mm (0.08 in.) thick
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
99.999%
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

RTECSNumber : MD5070000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents