missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011388FF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold foil | |
7440-57-5 | |
Au | |
25 x 25 mm | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
99.9985% | |
Odorless | |
Foil |
1.0mm (0.04 in.) thick | |
(metals basis) | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Premion™ | |
≈12g/25x25mm |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only