Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold foil, 1.0mm (0.04 in.) thick, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold foil
CAS 7440-57-5
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14723FF

  SDS
alfa aesar™
14723FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay Percent Notes (metals basis)
Quantity 25x25mm
Physical Form Foil
Name Note 1.0mm (0.04 in.) thick
Odor Odorless
CAS 7440-57-5
Weight ≈12g/25x25mm
Assay 99.95%
MDL Number MFCD00003436
Chemical Name or Material Gold foil
TSCA TSCA