Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold foil, 0.5mm (0.02 in.) thick, Premion™, 99.9985% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold foil
CAS 7440-57-5
Name Note 0.5mm (0.02 in.) thick
Purity Grade Premion™
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11389FF

  SDS
alfa aesar™
11389FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold foil
TSCA TSCA
Name Note 0.5mm (0.02 in.) thick
Purity Grade Premion™
Physical Form Foil
Assay 99.9985%
Assay Percent Notes (metals basis)
MDL Number MFCD00003436
CAS 7440-57-5
Odor Odorless
Weight ≈6g/25x25mm
Quantity 25x25mm