Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold foil, 0.5mm (0.02 in.) thick, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold foil
CAS 7440-57-5
Name Note 0.5mm (0.02 in.) thick
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36148FF

  SDS
alfa aesar™
36148FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Weight ≈6g/25x25mm
Chemical Name or Material Gold foil
Assay Percent Notes (metals basis)
Physical Form Foil
CAS 7440-57-5
Name Note 0.5mm (0.02 in.) thick
TSCA TSCA
MDL Number MFCD00003436
Assay 99.95%
Quantity 25x25mm
Odor Odorless