missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 1245306

 View more versions of this product

Catalog No. AA1245306



Specifications

Specifications

Gold flake/sphere
7440-57-5
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.967
CHEBI:30050
APS 1 to 15 Micron
0.24 to 0.34m2/g
5g
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Flakes or Spheres
99.95%

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents