Please login to your online account to display your discounted pricing

Gold Chloride, Trihydrate, ACS Reagent Grade, Ricca Chemical

$473.09 - $473.09

Chemical Identifiers

Chemical Name or Material Gold Chloride
Name Note Trihydrate
CAS 16961-25-4
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDCG05001C2

 
ricca chemical company
RIRDCG05001C2
Each for $473.09
Description & Specifications

Specifications

Chemical Name or Material Gold Chloride
Shelf Life 60 months
Name Note Trihydrate
CAS 16961-25-4
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Quantity 1mL
Safety and Handling

60 months