missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold Chloride, Trihydrate, ACS Grade, ≥ 49.0 % Au, LabChem™

$303.60 - $303.60

Chemical Identifiers

CAS 64-17-5,7732-18-5,67-63-0,67-56-1,9000-29-7
CAS Max % 84.1,4.92,4.67,4.67,1.64
Linear Formula HAuCl4·3H2O
Formula Weight 393.83
Chemical Name or Material Gold Chloride, Trihydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC148937 Lab Chem
LC148937
1g Each for $303.60
Specifications Description & Specifications

Chemical Identifiers

64-17-5,7732-18-5,67-63-0,67-56-1,9000-29-7
HAuCl4·3H2O
Gold Chloride, Trihydrate
84.1,4.92,4.67,4.67,1.64
393.83
ACS Grade

Specifications

64-17-5,7732-18-5,67-63-0,67-56-1,9000-29-7
Clump
393.83
Vial
Refrigerate
Gold
1g
≥49.0% Au
84.1,4.92,4.67,4.67,1.64
HAuCl4·3H2O
Passes Test
Soluble in water
Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
254°C
Gold Chloride, Trihydrate
ACS Grade

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger