Please login to your online account to display your discounted pricing

Gold Chloride, 1% (w/v), LabChem™

$647.69 - $647.69

Chemical Identifiers

Chemical Name or Material Gold Chloride
CAS 7732-18-5
CAS Max % 98.9
CAS 16961-25-4
CAS Max % 1.1
Linear Formula HAuCl4·3H2O
Formula Weight 393.83
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC148957

 
lab chem
LC148957
125mL Each for $647.69
Description & Specifications

Specifications

Formula Weight 393.83
Chemical Name or Material Gold Chloride
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Assay 1% (w/v)
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula HAuCl4·3H2O
CAS 7732-18-5
Decomposition Information Hydrogen chloride
Color Clear, yellow
Quantity 125mL
Health Hazard 1 Warning
CAS Max % 98.9
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 16961-25-4
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS Max % 1.1
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.