missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold Chloride, 1% (w/v) Aqueous Solution, 100mL = 1 g HAuCl·3H2O, Ricca Chemical
Toner For Gomori Methenamine Silver Nitrate Stain
Manufacturer: Ricca Chemical Company RI32954
Specifications
Gold Chloride | |
16961-25-4,7732-18-5 | |
100.0,1.01 | |
Amber Glass | |
100 mL = 1 g HAuCl·3H2O; 1mL = 5mg Au | |
120mL |
1% (w/v) | |
97.0,0.97 | |
Liquid | |
Aqueous Solution | |
Yellow |
Chemical Identifiers
16961-25-4 |