missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold, AAS standard solution, Specpure™, Au 1000μg/mL

Manufacturer:  Alfa Aesar™ 88068AE

 View more versions of this product

Catalog No. AA88068AE



Specifications

Specifications

Gold, AAS standard solution
Au in 20% HCl
UN1789
Specpure™
Au 1000μg/mL
MFCD00003436
Liquid
100mL

Chemical Identifiers

MFCD00003436
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID

TSCA : TSCA

SDS Documents