missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold AA Standard, Certified, 1000 ppm (1mL = 1mg Au), LabChem™

Manufacturer:  LabChem LC148907

 View more versions of this product

Catalog No. LC148907



Specifications

Specifications

Gold AA Standard
16961-25-4,7647-01-0,7732-18-5
HAuCl4·3H2O
UN3264
Liquid
Certified
Soluble in water
Yellow
1000ppm (1mL = 1mg Au)
89.82,9.98,0.2
1000 ppm (1mL = 1mg Au)
393.83
Passes Test
Amber Glass
Traceable to NIST
125mL

Chemical Identifiers

16961-25-4
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions