missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GOJO™ RICH PINK™ Antibacterial Lotion Soap

2000 mL Refill for GOJO™ PRO™ TDX™ Dispenser

Manufacturer:  GOJO™ 722004CAN

Catalog No. 19-077-515




Description

Description

  • High performance, antibacterial lotion soap for general handwashing
  • Quickly builds to a rich lather
  • Contains skin conditioners
  • Gentle on skin
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
Specifications

Specifications

0.52 gal.
Refill
Ethanolamine <3%, Oleic Acid, Sodium Sulfate
Antibacterial Lotion Soap
Floral balsam
7 to 10
2000mL
4 Case
Pink
Liquid
For cleaning light soils
PRO 2000 Dispenser
SDS
Documents
Product Certifications