missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Goat anti-Rat IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Manufacturer: Mp Biomedicals 0855778
Description
Host Species: Goat
Species Reactivity: Rat
Specifications
IgG, Whole Molecule | |
Lyophilized | |
2mL | |
Polyclonal |
HRP | |
Goat | |
Secondary | |
Rat |