Please login to your online account to display your discounted pricing

Goat anti-Rat Albumin Polyclonal Antibody, MP Biomedicals

Manufacturer: mp biomedicals  55727

 View all options for this product

Catalog No. ICN55727


Description & Specifications


Antigen Albumin
Conjugate Unlabeled
Cross Reactivity Rat
Format Lyophilized
Host Species Goat
Quantity 5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal

Host Species: Goat
Species Reactivity: Rat