missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Goat anti-Rat Albumin Polyclonal Antibody, MP Biomedicals
$269.22 - $269.22
Specifications
Antigen | Albumin |
---|---|
Format | Lyophilized |
Host Species | Goat |
Monoclonal or Polyclonal | Polyclonal |
Primary or Secondary | Primary |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN55710
|
Mp Biomedicals
0855710 |
2mL |
Each for $269.22
|
|
Description
Host Species: Goat
Species Reactivity: Rat
Specifications
Albumin | |
Goat | |
Primary |
Lyophilized | |
Polyclonal |