missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Goat anti-Rabbit IgG, Poly-HRP, Ampli-HRP, Polyclonal Secondary Antibody, MilliporeSigma™

$210.19 - $210.19

Specifications

Host Species Goat
Antigen Goat anti-Rabbit IgG
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Secondary
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AP342P7MLMI View Documents MilliporeSigma
AP342P7ML
7mL Each for $210.19
 
Specifications

Specifications

Goat
Polyclonal
Secondary
Liquid.
Goat anti-Rabbit IgG
Affinity Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)

For Research Use Only