Please login to your online account to display your discounted pricing

Goat anti-Rabbit IgG, Poly-HRP, Ampli-HRP, Polyclonal Secondary Antibody, EMD Millipore™


Manufacturer: emd millipore corporation  AP342P7ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AP342P7MLMI

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Rabbit
Quantity 7mL
Host Species Goat
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Goat anti-Rabbit IgG
Formulation Liquid.
Conjugate HRP
Storage Requirements 2-8°C for up to 12 months.
Primary or Secondary Secondary
Purification Method Immunoaffinity purified
Monoclonal or Polyclonal Polyclonal