missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
IgMμ Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: Emd Millipore Corporation 12489
Description
Goat anti-Mouse IgMμ Antibody, HRP conjugate detects level of Mouse IgMμ and has been published and validated for use in western blotting.Specifications
Goat anti-Mouse IgMμ | |
Polyclonal | |
0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/mL BSA, 0.01% Gentamicin Sulfate | |
250 μg | |
Mouse |
Western Blot | |
HRP | |
Goat | |
Secondary | |
−20°C for up to 12 months. |
For Research Use Only