Please login to your online account to display your discounted pricing

Goat anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals

Manufacturer: mp biomedicals  55435

 View all options for this product

Catalog No. ICN55435


Description & Specifications


Antigen IgG, Whole Molecule
Conjugate Unlabeled
Cross Reactivity Mouse
Format Lyophilized
Host Species Goat
Isotype IgG
Quantity 2mL
Primary or Secondary Secondary
Monoclonal or Polyclonal Polyclonal

Host Species: Goat
Isotype: IgG
Species Reactivity: Mouse