Please login to your online account to display your discounted pricing

Goat anti-Mouse IgG, Poly-HRP, Ampli-HRP, Polyclonal Secondary Antibody, EMD Millipore™


Manufacturer: emd millipore corporation  AP340P50ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AP340P50ML

  • / Each
 

Description & Specifications

Specifications

Host Species Goat
Quantity 50mL
Conjugate HRP
Antigen Goat anti-Mouse IgG
Format Affinity Purified
Formulation Liquid.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Purification Method Immunoaffinity purified
Storage Requirements 2-8°C for up to 12 months.
Primary or Secondary Secondary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Mouse