missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Goat anti-Mouse IgG, Poly-HRP, Ampli-HRP, Polyclonal Secondary Antibody, MilliporeSigma™

$708.31 - $708.31

Specifications

Host Species Goat
Antigen Goat anti-Mouse IgG
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Secondary
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AP340P50ML View Documents MilliporeSigma
AP340P50ML
50mL Each for $708.31
 
Specifications

Specifications

Goat
Polyclonal
Secondary
Liquid.
Goat anti-Mouse IgG
Affinity Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)

For Research Use Only