missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG γ chain Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Mouse IgG γ chain Antibody, Species Adsorbed detects level of Mouse IgG γ chain and has been published and validated for use in ELISA.
Specifications
Specifications
| Antigen | Goat anti-Mouse IgG γ chain Species adsorbed |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Liquid in 1.0mLof 100mM borate buffered saline, pH 8.2. |
| Host Species | Goat |
| Immunogen | purified mouse IgG γ chain. Pooled antisera from goats hyperimmunized with mouse IgG paraproteins. |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only