missing translation for 'onlineSavingsMsg'
Learn More

IgG γ chain Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP503PMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP503PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP503PMI Supplier MilliporeSigma Supplier No. AP503P
Only null left

Goat Polyclonal Antibody

Goat anti-Mouse IgG γ chain Antibody, HRP conjugate, Species Adsorbed is an antibody against Mouse IgG γ chain for use in ELISA.

Specifications

Antigen Goat anti-Mouse IgG γ chain Species adsorbed
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation Purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Liquid in 50% glycerol/50% PBS, pH 7.4.
Host Species Goat
Immunogen purified mouse IgG γ chain. Pooled antisera from goats hyperimmunized with mouse IgG paraproteins.
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months −20°C in undiluted aliquots for long term storage. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only