missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Goat anti-Human IgG Fc, γ-Specific Polyclonal Secondary Antibody, MP Biomedicals
Manufacturer: Mp Biomedicals 0855014
Description
Host Species: Goat
Species Reactivity: Human
Specifications
IgG Fc, gamma-specific | |
Purified | |
5mL | |
Polyclonal |
Unconjugated | |
Goat | |
Secondary | |
Human |