missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Goat anti-Human IgG Fc, γ-Specific Polyclonal Secondary Antibody, MP Biomedicals

$216.63 - $216.63

Specifications

Antigen IgG Fc, gamma-specific
Format Purified
Host Species Goat
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN55014
View Documents
Mp Biomedicals
0855014
5mL
Each for $216.63
 
Description

Description

Host Species: Goat
Species Reactivity: Human

Specifications

Specifications

IgG Fc, gamma-specific
Goat
Secondary
Purified
Polyclonal
SDS