Please login to your online account to display your discounted pricing

Goat anti-Human IgG Fc, γ-Specific Polyclonal Secondary Antibody, MP Biomedicals

Manufacturer: mp biomedicals  55095

 View all options for this product

Catalog No. ICN55095


Description & Specifications


Antigen IgG Fc, gamma-Specific
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Host Species Goat
Quantity 2mL
Primary or Secondary Secondary
Monoclonal or Polyclonal Polyclonal

Host Species: Goat
Species Reactivity: Human