missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Goat anti-Human IgA, alpha-Specific Polyclonal Secondary Antibody, MP Biomedicals
Manufacturer: Mp Biomedicals 0855082
Description
Host Species: Goat
Species Reactivity: Human
Specifications
IgA, alpha-Specific | |
Purified | |
2mL | |
Polyclonal |
Unconjugated | |
Goat | |
Secondary |