missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Goat anti-Human IgA, alpha-Specific Polyclonal Secondary Antibody, MP Biomedicals
$119.72 - $119.72
Specifications
Antigen | IgA, alpha-Specific |
---|---|
Format | Purified |
Host Species | Goat |
Monoclonal or Polyclonal | Polyclonal |
Primary or Secondary | Secondary |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN55082
|
Mp Biomedicals
0855082 |
2mL |
Each for $119.72
|
|
Description
Host Species: Goat
Species Reactivity: Human
Specifications
IgA, alpha-Specific | |
Goat | |
Secondary |
Purified | |
Polyclonal |