missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig λ chain Goat anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Human Ig λ chain Antibody, HRP conjugate detects level of Human Ig λ chain and has been published and validated for use in ELISA.
Specifications
Specifications
| Antigen | Goat anti-Human Ig λ chain |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 50% glycerol/50% PBS, pH 7.4. |
| Host Species | Goat |
| Immunogen | pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains. |
| Purification Method | Affinity Purified |
| Quantity | 1 mL |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only