missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig κ chain Goat anti-Human, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Human Ig κ chain Antibody, FITC conjugate is validated for use in IF for the detection of Human Ig κ chain.
Specifications
Specifications
| Antigen | Goat anti-Human Ig κ chain |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | Purified by affinity chromatography on pooled human Igs with kappa light chains covalently linked to agarose. Liquid in 1.0mLPBS/NaN3. |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Show More |
For Research Use Only