missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (H+L) Goat anti-Chicken, Phycoerythrin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Chicken IgG (H+L) Antibody, PE conjugate is an antibody against Chicken IgG (H+L) for use in ELISA and immunofluoresence.
Specifications
Specifications
| Antigen | Goat anti-Chicken IgG (HL) |
| Applications | ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | PE |
| Formulation | Purified by affinity chromatography on chicken IgG covalently linked to agarose. Liquid in 1.0mLof PBS/NaN3 and a stabilizing agent. |
| Host Species | Goat |
| Immunogen | pooled antisera from goats hyperimmunized with chicken IgG. |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Show More |
For Research Use Only