Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

$277.82 - $1,198.12

Specifications

Antigen GnRH Receptor / LH-RH Receptor Ab-3
Clone GNRH03; same as F1G4
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1139P

 
thermo scientific™ lab vision™
MS1139P
Purified Ab with BSA and Azide 1mL Each for $1,123.47

MS1139P0

 
thermo scientific™ lab vision™
MS1139P0
Purified Ab with BSA and Azide 100μL Each for $277.82

MS1139P1

 
thermo scientific™ lab vision™
MS1139P1
Purified Ab with BSA and Azide 500μL Each for $745.57

MS1139P1ABX

 
thermo scientific™ lab vision™
MS1139P1ABX
Ab wWithout BSA/Azide (Frozen) 100μL Each for $820.22

MS1139PABX

 
thermo scientific™ lab vision™
MS1139PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,198.12

MS1139R7

 
thermo scientific™ lab vision™
MS1139R7
Ready-to-use for IHC (UVLP) 7mL Each for $672.86
Description & Specifications

Specifications

Antigen GnRH Receptor / LH-RH Receptor Ab-3
Clone GNRH03; same as F1G4
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline G Protein-Coupled Receptors
Type Primary
Immunogen A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor
Isotype IgG1

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody.

GnRH receptor contains seven hydrophobic transmembrane domains connected by hydrophilic extracellular and intracellular loops characteristic of G-protein coupled receptors. GnRH stimulates the gonadotrophs of the anterior pituitary to secrete luteinizing hormone (LH) as well as follicle-stimulating hormone (FSH).

Host Species: Mouse

Clone: GNRH03; same as F1G4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 1-29

Immunogen: A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor

Molecular Weight: 54-60 kDa

Positive Control: Pituitary or normal ovary

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO