Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS1139PABX

 View more versions of this product

Catalog No. MS1139PABX

  • / Each
 

Description & Specifications

Specifications

Antigen GnRH Receptor / LH-RH Receptor Ab-3
Format Concentrated
Quantity 200µL
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Isotype IgG1
Clone GNRH03; same as F1G4
Host Species Mouse
Immunogen A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor
Regulatory Status RUO
Cross Reactivity Human
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline G Protein-Coupled Receptors
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody.

GnRH receptor contains seven hydrophobic transmembrane domains connected by hydrophilic extracellular and intracellular loops characteristic of G-protein coupled receptors. GnRH stimulates the gonadotrophs of the anterior pituitary to secrete luteinizing hormone (LH) as well as follicle-stimulating hormone (FSH).

Host Species: Mouse

Clone: GNRH03; same as F1G4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 1-29

Immunogen: A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor

Molecular Weight: 54-60 kDa

Positive Control: Pituitary or normal ovary

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO