Please login to your online account to display your discounted pricing

GNF 5837, Tocris™

Potent Trk inhibitor; inhibits TrkA, TrkB and TrkC

$310.33 - $1,282.40

Chemical Identifiers

Chemical Name or Material GNF 5837
Assay Percent Range >98%
Molecular Formula C28H21F4N5O2
Formula Weight 535.49
Synonym N-[3-[[2, 3-Dihydro-2-oxo-3-(1H-pyrrol-2-ylmethylene)-1H-indol-6-yl]amino]-4-methylphenyl]-N'-[2-fluoro-5-(trifluoromethyl)phenyl]urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

455950

 
r & d systems
455950
50mg Each for $1,282.40

455910

 
r & d systems
455910
10mg Each for $310.33
Description & Specifications

Specifications

Formula Weight 535.49
Quantity 50mg
Chemical Name or Material GNF 5837
Assay Percent Range >98%
Molecular Formula C28H21F4N5O2
CAS 1033769-28-6
Synonym N-[3-[[2, 3-Dihydro-2-oxo-3-(1H-pyrrol-2-ylmethylene)-1H-indol-6-yl]amino]-4-methylphenyl]-N'-[2-fluoro-5-(trifluoromethyl)phenyl]urea
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C