Please login to your online account to display your discounted pricing

GNF 2, Tocris™

Selective allosteric inhibitor of Bcr-Abl tyrosine kinase activity

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material GNF 2
Assay Percent Range >98%
Molecular Formula C18H13F3N4O2
Formula Weight 374.32
Synonym 3-[6-[[4-(Trifluoromethoxy)phenyl]amino]-4-pyrimidinyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439910

 
r & d systems
439910
10mg Each for $284.81

439950

 
r & d systems
439950
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 374.32
Quantity 10mg
Chemical Name or Material GNF 2
Molecular Formula C18H13F3N4O2
CAS 778270-11-4
Synonym 3-[6-[[4-(Trifluoromethoxy)phenyl]amino]-4-pyrimidinyl]benzamide
Assay Percent Range >98%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C