Please login to your online account to display your discounted pricing

Tocris™ GMQ hydrochloride

ASIC3 channel opener

$102.98 - $436.69

Specifications

Product Type GMQ hydrochloride
Inhibitors GMQ hydrochloride
Molecular Weight 237.69
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

502810R

 
tocris™
502810
10mg Each for $102.98

502850R

 
tocris™
502850
50mg Each for $436.69
Description & Specifications

Specifications

Product Type GMQ hydrochloride
Inhibitors GMQ hydrochloride
Molecular Weight 237.69
Purity >98

Selective opener of acid-sensing ion channel 3 (ASIC3) at pH 7.4. Alters the pH dependence of ASIC1a and ASIC1b; does not affect the activation curve of ASIC2a.

  • Chemical Name: N-(4-Methyl-2-quinazolinyl)-guanidine hydrochloride
  • Formula: C10H11N5.HCl
  • CAS Number: 5361-15-9
  • Solubility: Soluble to 50mM in DMSO and to 5mM in water with gentle warming
  • Purity: >98