Please login to your online account to display your discounted pricing

GM-CSF Rat anti-Mouse, PE, Clone: MP1-22E9, eBioscience™

Rat Monoclonal Antibody

$158.05 - $317.55

Specifications

Antigen GM-CSF
Clone MP1-22E9
Host Species Rat
Gene Alias Granulocyte-macrophage colony stimulating factor, CSF2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011196

 
life technologies
LS12733181
50μg Each for $220.40

501122387

 
life technologies
LS12733141
25 Tests Each for $158.05

501122388

 
life technologies
LS12733182
100μg Each for $317.55
Description & Specifications

Specifications

Antigen GM-CSF
Clone MP1-22E9
Host Species Rat
Gene Alias Granulocyte-macrophage colony stimulating factor, CSF2
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The MP1-22E9 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF). The MP1-22E9 antibody is a neutralizing antibody. Mouse GM-CSF is a 14kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF-α, IL-2, IL-1, and IFN-γ. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active.