Please login to your online account to display your discounted pricing

GM-CSF Rat anti-Mouse, Biotin, Clone: MP1-31G6, eBioscience™

Rat Monoclonal Antibody

$143.55 - $404.55

Specifications

Antigen GM-CSF
Clone MP1-31G6
Host Species Rat
Gene Alias Granulocyte-macrophage colony stimulating factor, GMCSF, Colony-stimulating factor, CSF12
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012207

 
life technologies
LS13733281
50μg Each for $143.55

5012208

 
life technologies
LS13733285
500μg Each for $404.55
Description & Specifications

Specifications

Antigen GM-CSF
Clone MP1-31G6
Host Species Rat
Gene Alias Granulocyte-macrophage colony stimulating factor, GMCSF, Colony-stimulating factor, CSF12
Species Reactivity Mouse
Applications ELISA Detection
Applications ELISPOT Detection Antibody
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MP1-31G6 antibody reacts with mouse granulocyte/macrophage - colony stimulating factor (GM-CSF), a 14kDa cytokine secreted by activated T cells and macrophages.