Please login to your online account to display your discounted pricing

GM-CSF Rat anti-Human, PE, Clone: DAVKAT, eBioscience™

Rat Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen GM-CSF
Clone DAVKAT
Host Species Rat
Gene Alias Granulocyte-Macrophage Colony-Stimulating factor, CSF2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122386

 
life technologies
LS12720941
25 Tests Each for $172.55

501129344

 
life technologies
LS12720942
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen GM-CSF
Clone DAVKAT
Host Species Rat
Gene Alias Granulocyte-Macrophage Colony-Stimulating factor, CSF2
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This DAVKAT monoclonal antibody reacts with human Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF). Human GM-CSF is a 127 amino acid, differentially glycosylated factor produced mainly by activated T cells and macrophages. Endothelial cells and fibroblasts can also produce GM-CSF after exposure to TNF alpha, IL-1, IL-2 and IFN gamma. GM-CSF is found associated with the extracellular matrix and in membrane-bound formats as well. It stimulates proliferation, activation and differentiation of macrophages, granulocytes and their progenitors.